5-(4-Chloro-3-methylphenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

55374-1G 55374-500MG 55374-5G
76933-754EA 494.38 USD
76933-754 76933-756 76933-758
5-(4-Chloro-3-methylphenyl)thiazol-2-amine ≥95%
5-(4-Chloro-3-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂S
MW: 224.71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12022356
CAS Number: 1195901-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR