5-(3-Bromo-5-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55313-1G 55313-25G 55313-500MG 55313-5G
76933-724EA 363.89 USD
76933-724 76933-726 76933-728 76933-730
5-(3-Bromo-5-fluorophenyl)thiazol-2-amine ≥97%
5-(3-Bromo-5-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆BrFN₂S
MW: 273.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29481182
CAS Number: 1936670-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR