5-(3,5-Difluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55254-1G 55254-25G 55254-500MG 55254-5G
76933-684EA 331.01 USD
76933-684 76933-686 76933-688 76933-690
5-(3,5-Difluorophenyl)thiazol-2-amine ≥97%
5-(3,5-Difluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆F₂N₂S
MW: 212.219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12824047
CAS Number: 1183990-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR