5-(5-Bromo-2-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55168-1G 55168-500MG 55168-5G
76933-672EA 500.75 USD
76933-672 76933-674 76933-676
5-(5-Bromo-2-methylphenyl)thiazol-2-amine ≥97%
5-(5-Bromo-2-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂S
MW: 269.161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530361
CAS Number: 2734773-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR