5-(3-Fluoro-5-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55158-1G 55158-500MG 55158-5G
76933-646EA 560.16 USD
76933-646 76933-648 76933-650
5-(3-Fluoro-5-methylphenyl)thiazol-2-amine ≥97%
5-(3-Fluoro-5-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉FN₂S
MW: 208.255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23750651
CAS Number: 1542102-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR