5-(2,4-Dimethylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55141-1G 55141-500MG 55141-5G
76933-620EA 464.68 USD
76933-620 76933-622 76933-624
5-(2,4-Dimethylphenyl)thiazol-2-amine ≥97%
5-(2,4-Dimethylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂S
MW: 204.291 g/mol
Storage Temperature: Ambient
CAS Number: 1182997-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR