5-Chloro-2-fluoro-4-(trifluoromethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

55120-1G 55120-25G 55120-500MG 55120-5G
76933-590EA 393.6 USD
76933-590 76933-592 76933-594 76933-596
5-Chloro-2-fluoro-4-(trifluoromethyl)benzaldehyde ≥95%
5-Chloro-2-fluoro-4-(trifluoromethyl)benzaldehyde
Formula: C₈H₃ClF₄O
MW: 226.55543 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32708897
CAS Number: 134099-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR