4-Iodo-2-methylphenol ≥97%

Supplier: AOBChem USA

Warning

55100-25G 55100-5G
76933-578EA 325.7 USD
76933-578 76933-580
4-Iodo-2-methylphenol ≥97%
4-Iodo-2-methylphenol
Formula: C₇H₇IO
MW: 234.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075092
CAS Number: 60577-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR