5-(2-Ethylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55069-1G 55069-500MG 55069-5G
76933-550EA 500.75 USD
76933-550 76933-552 76933-554
5-(2-Ethylphenyl)thiazol-2-amine ≥97%
5-(2-Ethylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂S
MW: 204.291 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530353
CAS Number: 1595674-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR