5-(4-(Methylthio)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55060-1G 55060-25G 55060-500MG 55060-5G
76933-534EA 266.29 USD
76933-534 76933-536 76933-538 76933-540
5-(4-(Methylthio)phenyl)thiazol-2-amine ≥97%
5-(4-(Methylthio)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂S₂
Storage Temperature: Ambient
CAS Number: 1823409-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR