5-(3-Chlorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

54990-1G 54990-25G 54990-500MG 54990-5G
76933-512EA 363.89 USD
76933-512 76933-514 76933-516 76933-518
5-(3-Chlorophenyl)thiazol-2-amine ≥97%
5-(3-Chlorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₇ClN₂S
MW: 210.683 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14600376
CAS Number: 1249447-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR