5-(3-Ethylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

54984-1G 54984-25G 54984-500MG 54984-5G
76933-498EA 381.93 USD
76933-498 76933-500 76933-502 76933-504
5-(3-Ethylphenyl)thiazol-2-amine ≥97%
5-(3-Ethylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂S
MW: 204.291 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24127585
CAS Number: 1509544-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR