5-(3,5-Dimethylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

54979-1G 54979-25G 54979-500MG 54979-5G
76933-484EA 428.61 USD
76933-484 76933-486 76933-488 76933-490
5-(3,5-Dimethylphenyl)thiazol-2-amine ≥97%
5-(3,5-Dimethylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂S
MW: 204.291 g/mol
Storage Temperature: Ambient
CAS Number: 1183989-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR