5-(4-Ethylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

54975-1G 54975-25G 54975-500MG 54975-5G
76933-470EA 441.34 USD
76933-470 76933-472 76933-474 76933-476
5-(4-Ethylphenyl)thiazol-2-amine ≥97%
5-(4-Ethylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂S
MW: 204.291 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12779913
CAS Number: 73040-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR