5-Bromo-3-(trifluoromethoxy)salicylaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-hydroxy-3-(trifluoromethoxy)benzaldehyde

54866-1G 54866-25G 54866-5G
76933-460EA 84.88 USD
76933-460 76933-462 76933-464
5-Bromo-3-(trifluoromethoxy)salicylaldehyde ≥97%
5-Bromo-3-(trifluoromethoxy)salicylaldehyde

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₃
MW: 285.02
Storage Temperature: Refrigerator
MDL Number: MFCD03788532
CAS Number: 497959-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR