5-(4-(Trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

54850-1G 54850-25G 54850-500MG 54850-5G
76933-446EA 434.97 USD
76933-446 76933-448 76933-450 76933-452
5-(4-(Trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(4-(Trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₇F₃N₂S
MW: 244.236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12779867
CAS Number: 1178075-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR