5-(4-Fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

54848-1G 54848-25G 54848-500MG 54848-5G
76933-430EA 391.48 USD
76933-430 76933-432 76933-434 76933-436
5-(4-Fluorophenyl)thiazol-2-amine ≥97%
5-(4-Fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₇FN₂S
MW: 194.229 g/mol
Storage Temperature: Ambient
MDL Number: MFCD5864559
CAS Number: 774-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR