5-((tert-Butyldimethylsilyl)oxy)-2-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

54515-1G 54515-25G 54515-500MG 54515-5G
76933-404EA 203.7 USD
76933-404 76933-406 76933-408 76933-410
5-((tert-Butyldimethylsilyl)oxy)-2-fluorobenzoic acid ≥95%
5-((tert-Butyldimethylsilyl)oxy)-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₉FO₃Si
MW: 270.372 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34530269
CAS Number: 179117-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR