5-(3-Bromo-4-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

54300-1G 54300-25G 54300-500MG 54300-5G
76933-376EA 381.93 USD
76933-376 76933-378 76933-380 76933-382
5-(3-Bromo-4-fluorophenyl)thiazol-2-amine ≥97%
5-(3-Bromo-4-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆BrFN₂S
MW: 273.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29932075
CAS Number: 1592317-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR