4-Iodo-2,6-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53874-1G 53874-25G 53874-5G
76933-326EA 84.88 USD
76933-326 76933-328 76933-330
4-Iodo-2,6-dimethylbenzaldehyde ≥97%
4-Iodo-2,6-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉IO
Storage Temperature: Refrigerator
MDL Number: MFCD34178714
CAS Number: 160682-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR