5-Chloro-2-methoxy-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53642-1G 53642-500MG 53642-5G
76933-312EA 545.31 USD
76933-312 76933-314 76933-316
5-Chloro-2-methoxy-4-methylbenzaldehyde ≥97%
5-Chloro-2-methoxy-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Storage Temperature: Ambient
CAS Number: 82128-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR