5-Bromo-2-chloro-6-methylnicotinic acid ≥97%

Supplier: AOBChem USA

Warning

53534-1G 53534-25G 53534-500MG 53534-5G
76933-298EA 233.4 USD
76933-298 76933-300 76933-302 76933-304
5-Bromo-2-chloro-6-methylnicotinic acid ≥97%
5-Bromo-2-chloro-6-methylnicotinic acid
Formula: C₇H₅BrClNO₂
MW: 250.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18258584
CAS Number: 1256809-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR