5-Bromo-2-fluoro-4-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

53277-1G 53277-25G 53277-500MG 53277-5G
76933-256EA 274.78 USD
76933-256 76933-258 76933-260 76933-262
5-Bromo-2-fluoro-4-(trifluoromethyl)benzoic acid ≥97%
5-Bromo-2-fluoro-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₃BrF₄O₂
MW: 287.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29691075
CAS Number: 1699741-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR