4-Chloro-2-fluoro-5-nitrobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53113-1G 53113-25G 53113-500MG 53113-5G
76933-240EA 153.84 USD
76933-240 76933-242 76933-244 76933-246
4-Chloro-2-fluoro-5-nitrobenzaldehyde ≥97%
4-Chloro-2-fluoro-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClFNO₃
MW: 203.555 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28059711
CAS Number: 1639298-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR