4-Isobutylphenol ≥97%

Supplier: AOBChem USA

Warning

53022-1G 53022-25G 53022-500MG 53022-5G
76933-220EA 233.4 USD
76933-220 76933-222 76933-224 76933-226
4-Isobutylphenol ≥97%
4-Isobutylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD01846454
CAS Number: 4167-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR