5-Bromo-2-((4-methoxybenzyl)oxy)pyrimidine ≥95%

Supplier: AOBChem USA

Warning

52962-1G 52962-25G 52962-5G
76933-206EA 117.76 USD
76933-206 76933-208 76933-210
5-Bromo-2-((4-methoxybenzyl)oxy)pyrimidine ≥95%
5-Bromo-2-((4-methoxybenzyl)oxy)pyrimidine
Formula: C₁₂H₁₁BrN₂O₂
MW: 295.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12026110
CAS Number: 1159000-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR