4-Bromo-2-fluorophenyl methyl sulfone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-fluoro-1-(methylsulphonyl)benzene

52869-1G 52869-25G 52869-5G
76933-192EA 76.39 USD
76933-192 76933-194 76933-196
4-Bromo-2-fluorophenyl methyl sulfone ≥95%
4-Bromo-2-fluorophenyl methyl sulfone
Formula: C₇H₆BrFO₂S
MW: 253.089 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11855948
CAS Number: 648904-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR