5-Formyl-2-methylbenzonitrile ≥97%

Supplier: AOBChem USA

Warning

52726-1G 52726-25G 52726-5G
76933-180EA 63.66 USD
76933-180 76933-182 76933-184
5-Formyl-2-methylbenzonitrile ≥97%
5-Formyl-2-methylbenzonitrile
Formula: C₉H₇NO
Storage Temperature: Ambient
MDL Number: MFCD12197728
CAS Number: 27613-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR