4-Cyano-3-methylbenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

52589-25G 52589-5G
76933-156EA 233.4 USD
76933-156 76933-158
4-Cyano-3-methylbenzoic acid ≥97% research grade
4-Cyano-3-methylbenzoic acid
Formula: C₉H₇NO₂
MW: 161.1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14582961
CAS Number: 73831-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR