4-Iodo-2,5-dimethylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

52560-1G 52560-25G 52560-500MG 52560-5G
76933-142EA 441.34 USD
76933-142 76933-144 76933-146 76933-148
4-Iodo-2,5-dimethylbenzaldehyde ≥95%
4-Iodo-2,5-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉IO
MW: 260.072 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31381396
CAS Number: 2126161-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR