5-(tert-Butyl)-2-chlorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

52447-1G 52447-25G 52447-500MG 52447-5G
76933-126EA 263.11 USD
76933-126 76933-128 76933-130 76933-132
5-(tert-Butyl)-2-chlorobenzoic acid ≥95%
5-(tert-Butyl)-2-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₂
MW: 212.673 g/mol
Storage Temperature: Ambient
CAS Number: 60772-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR