4-Butoxy-3-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52282-1G 52282-25G 52282-500MG 52282-5G
76933-110EA 292.81 USD
76933-110 76933-112 76933-114 76933-116
4-Butoxy-3-fluorobenzoic acid ≥97%
4-Butoxy-3-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD01830206
CAS Number: 326-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR