5-Chloro-2-fluoro-3-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

51975-1G 51975-500MG 51975-5G
76933-076EA 587.74 USD
76933-076 76933-078 76933-080
5-Chloro-2-fluoro-3-formylbenzoic acid ≥97%
5-Chloro-2-fluoro-3-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClFO₃
MW: 202.567 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32902985
CAS Number: 2166733-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR