4-Fluoro-2-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

51697-1G 51697-25G 51697-500MG 51697-5G
76933-066EA 196.27 USD
76933-066 76933-068 76933-070 76933-072
4-Fluoro-2-methoxy-N-methylbenzamide ≥97%
4-Fluoro-2-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MW: 183.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403988
CAS Number: 2853499-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR