4-Phenylthiophene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

51688-25G 51688-5G
76933-056EA 573.95 USD
76933-056 76933-058
4-Phenylthiophene-2-carboxylic acid ≥97%
4-Phenylthiophene-2-carboxylic acid
Formula: C₁₁H₈O₂S
MW: 204.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03408007
CAS Number: 21676-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR