4-Fluoro-2-methoxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

51530-1G 51530-500MG 51530-5G
76933-046EA 554.86 USD
76933-046 76933-048 76933-050
4-Fluoro-2-methoxy-5-methylbenzaldehyde ≥97%
4-Fluoro-2-methoxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD31579900
CAS Number: 139008-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR