4-Chloro-3-ethoxy-2-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

51272-25G 51272-5G
76933-020EA 488.02 USD
76933-020 76933-022
4-Chloro-3-ethoxy-2-fluorobenzoic acid ≥95%
4-Chloro-3-ethoxy-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD19687215
CAS Number: 1323966-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR