4-Fluoro-3-formyl-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

50766-1G 50766-25G 50766-500MG 50766-5G
76932-984EA 263.11 USD
76932-984 76932-986 76932-988 76932-990
4-Fluoro-3-formyl-5-methylbenzoic acid ≥97%
4-Fluoro-3-formyl-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD34530059
CAS Number: 2384149-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR