5-Bromo-2,4-dichlorophenol ≥97% research grade

Supplier: AOBChem USA

Warning

50156-1G 50156-25G 50156-500MG 50156-5G
76932-968EA 154.9 USD
76932-968 76932-970 76932-972 76932-974
5-Bromo-2,4-dichlorophenol ≥97% research grade
5-Bromo-2,4-dichlorophenol
Formula: C₆H₃BrCl₂O
Storage Temperature: Refrigerator
MDL Number: MFCD16038723
CAS Number: 183803-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR