5-Iodo-2-methylanisole ≥95%

Supplier: AOBChem USA

Warning

49743-1G 49743-25G 49743-500MG 49743-5G
76932-952EA 173.99 USD
76932-952 76932-954 76932-956 76932-958
5-Iodo-2-methylanisole ≥95%
5-Iodo-2-methylanisole
Formula: C₈H₉IO
Storage Temperature: Ambient
MDL Number: MFCD06797971
CAS Number: 220728-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR