5-Bromo-4-fluoro-2-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-carboxy-5-fluorophenol, 5-Bromo-4-fluorosalicylic acid

48843-1G 48843-25G 48843-500MG 48843-5G
76932-840EA 151.71 USD
76932-840 76932-842 76932-844 76932-846
5-Bromo-4-fluoro-2-hydroxybenzoic acid ≥97%
5-Bromo-4-fluoro-2-hydroxybenzoic acid
Formula: C₇H₄BrFO₃
MW: 235.007 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08235149
CAS Number: 1644-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR