5-Bromo-4-ethylnicotinic acid ≥95%

Supplier: AOBChem USA

Warning

48574-1G 48574-25G 48574-500MG 48574-5G
76932-820EA 292.81 USD
76932-820 76932-822 76932-824 76932-826
5-Bromo-4-ethylnicotinic acid ≥95%
5-Bromo-4-ethylnicotinic acid

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD30722568
CAS Number: 2167106-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR