5-Bromo-2-iodo-1-methoxy-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

48494-1G 48494-25G 48494-500MG 48494-5G
76932-804EA 272.66 USD
76932-804 76932-806 76932-808 76932-810
5-Bromo-2-iodo-1-methoxy-3-methylbenzene ≥95%
5-Bromo-2-iodo-1-methoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrIO
MW: 326.957 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31672854
CAS Number: 1887769-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR