5-Bromo-2-chloroisonicotinonitrile ≥95%

Supplier: AOBChem USA

Warning

48209-1G 48209-25G 48209-500MG 48209-5G
76932-766EA 355.41 USD
76932-766 76932-768 76932-770 76932-772
5-Bromo-2-chloroisonicotinonitrile ≥95%
5-Bromo-2-chloroisonicotinonitrile
Formula: C₆H₂BrClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD16622809
CAS Number: 1211515-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR