5-(2,3-Dichlorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

47314-1G 47314-25G 47314-500MG 47314-5G
76932-710EA 369.2 USD
76932-710 76932-712 76932-714 76932-716
5-(2,3-Dichlorophenyl)thiazol-2-amine ≥97%
5-(2,3-Dichlorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆Cl₂N₂S
MW: 245.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12779843
CAS Number: 1216237-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR