5-(4-Bromophenyl)-3-(trifluoromethyl)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

46792-25G 46792-5G
76932-678EA 346.92 USD
76932-678 76932-680
5-(4-Bromophenyl)-3-(trifluoromethyl)-1H-pyrazole ≥97%
5-(4-Bromophenyl)-3-(trifluoromethyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₀H₆BrF₃N₂
MW: 291.067 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01554560
CAS Number: 219986-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR