4-Formyl-3-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

46408-1G 46408-25G 46408-500MG 46408-5G
76932-648EA 292.81 USD
76932-648 76932-650 76932-652 76932-654
4-Formyl-3-methoxybenzoic acid ≥97%
4-Formyl-3-methoxybenzoic acid
Formula: C₉H₈O₄
MW: 180.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20645589
CAS Number: 80893-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR