4-Bromo-2-propoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

46288-1G 46288-25G 46288-500MG 46288-5G
76932-634EA 155.96 USD
76932-634 76932-636 76932-638 76932-640
4-Bromo-2-propoxybenzaldehyde ≥97%
4-Bromo-2-propoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.097 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11619319
CAS Number: 1094555-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR