4-Methoxy-3-(methylthio)benzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

46178-1G 46178-500MG 46178-5G
76932-606EA 587.74 USD
76932-606 76932-608 76932-610
4-Methoxy-3-(methylthio)benzoic acid ≥97% research grade
4-Methoxy-3-(methylthio)benzoic acid
Formula: C₉H₁₀O₃S
Storage Temperature: Refrigerator
MDL Number: MFCD20640312
CAS Number: 87346-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR