5-Ethoxy-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

46142-1G 46142-25G 46142-500MG 46142-5G
76932-590EA 379.81 USD
76932-590 76932-592 76932-594 76932-596
5-Ethoxy-2-fluorobenzoic acid ≥97%
5-Ethoxy-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD32707160
CAS Number: 1936598-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR